4-(3-Chlorophenyl)thiazol-2-amine 95%

Supplier: Ambeed
Warning

A235585-100MG A235585-1G A235585-5G
77261-638EA 24.72 USD
77261-638 77261-640 77261-642
4-(3-Chlorophenyl)thiazol-2-amine 95%
4-(3-Chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂S
MW: 210.68 g/mol
MDL Number: MFCD02664012
CAS Number: 90533-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR