(2-Bromophenyl)(2,4-dimethylphenyl)sulfane 97%

Supplier: Ambeed
Warning

A235581-10G A235581-1G A235581-5G
77249-634EA 149.35 USD
77249-634 77249-636 77249-638
(2-Bromophenyl)(2,4-dimethylphenyl)sulfane 97%
(2-Bromophenyl)(2,4-dimethylphenyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrS
MW: 293.22 g/mol
MDL Number: MFCD26386449
CAS Number: 960203-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR