(3,4-Dichlorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dichlorophenylacetic acid

A235512-100G A235512-10G A235512-500G A235512-5G A235512-25G
77237-984EA 24.72 USD
77237-984 77237-986 77237-988 77237-990 77304-016
(3,4-Dichlorophenyl)acetic acid 97%
(3,4-Dichlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
MDL Number: MFCD00004333
CAS Number: 5807-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR