3-Hydroxy-4-methoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

A235484-100G A235484-10G A235484-25G A235484-5G
77275-866EA 85 USD
77275-866 77275-868 77275-870 77275-872
3-Hydroxy-4-methoxybenzyl alcohol 98%
3-Hydroxy-4-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MDL Number: MFCD00004644
CAS Number: 4383-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR