DL-Alanine methyl ester hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-DL-Ala-OMe.HCl, H-DL-Ala-OMe·HCl
A235480-100G
A235480-10G
A235480-25G
A235480-500G
A235480-5G
A235480-1000G
77275-856EA
31
USD
77275-856
77275-858
77275-860
77275-862
77275-864
77748-408
DL-Alanine methyl ester hydrochloride 98%
DL-Alanine methyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₉NO₂ MW: 139.58 g/mol |
MDL Number:
MFCD00035523 CAS Number: 13515-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...