5,11-Diphenyl-5,11-dihydroindolo[3,2-b]carbazole 97%

Supplier: Ambeed
Warning

A235472-1G A235472-250MG
77237-980EA 42.23 USD
77237-980 77237-982
5,11-Diphenyl-5,11-dihydroindolo[3,2-b]carbazole 97%
5,11-Diphenyl-5,11-dihydroindolo[3,2-b]carbazole

Precaución: for Research Use Only

Formula: C₃₀H₂₀N₂
MW: 408.49 g/mol
CAS Number: 58328-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR