(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl) ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-N,N'-(Cyclohexane-1,2-diyl)bis(2-(diphenylphosphino)benzamide), (R,R)-DACH-Phenyl Trost Ligand

A235469-100MG A235469-1G A235469-250MG A235469-5G A235469-25G
77237-972EA 14.42 USD
77237-972 77237-974 77237-976 77237-978 77550-516
(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl) ≥98%, ee 99%
(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N'-bis(2'-diphenylphosphinobenzoyl)

Precaución: for Research Use Only

Formula: C₄₄H₄₀N₂O₂P₂
MW: 690.7481 g/mol
MDL Number: MFCD01631273
CAS Number: 138517-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR