3,5-Dichloro-α,α,α-trifluorotoluene 97%

Supplier: Ambeed
Danger

A235314-1G A235314-250MG A235314-5G
77275-850EA 6.18 USD
77275-850 77275-852 77275-854
3,5-Dichloro-α,α,α-trifluorotoluene 97%
3,5-Dichloro-α,α,α-trifluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
MDL Number: MFCD00203929
CAS Number: 54773-20-5
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR