((1R,2S,4R)-4-((5-(1-(3-Bromobenzyl)-1H-pyrazole-3-carbonyl)pyrimidin-4-yl)amino)-2-hydroxycyclopentyl)methyl sulfamate 98%

Supplier: Ambeed
Warning

A235283-100MG A235283-10MG A235283-25MG A235283-50MG A235283-5MG A235283-250MG
77275-840EA 499 USD
77275-840 77275-842 77275-844 77275-846 77275-848 77748-407
((1R,2S,4R)-4-((5-(1-(3-Bromobenzyl)-1H-pyrazole-3-carbonyl)pyrimidin-4-yl)amino)-2-hydroxycyclopentyl)methyl sulfamate 98%
((1R,2S,4R)-4-((5-(1-(3-Bromobenzyl)-1H-pyrazole-3-carbonyl)pyrimidin-4-yl)amino)-2-hydroxycyclopentyl)methyl sulfamate

Precaución: for Research Use Only

Formula: C₂₁H₂₃BrN₆O₅S
MDL Number: MFCD32174257
CAS Number: 1644342-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR