[2,2'-Bipyridine]-4,4'-dicarbonitrile 97%

Supplier: Ambeed
Warning

A234549-100MG A234549-1G A234549-250MG A234549-5G
77288-518EA 36 USD
77288-518 77288-520 77288-522 77288-524
[2,2'-Bipyridine]-4,4'-dicarbonitrile 97%
[2,2'-Bipyridine]-4,4'-dicarbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₆N₄
MDL Number: MFCD06637688
CAS Number: 67491-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR