4-Fluorobenzaldehyde oxime 98%

Supplier: Ambeed
Danger

A234346-1G A234346-25G A234346-5G
77288-504EA 29 USD
77288-504 77288-506 77288-508
4-Fluorobenzaldehyde oxime 98%
4-Fluorobenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₇H₆FNO
MDL Number: MFCD00133101
CAS Number: 459-23-4
UN: 2926
ADR: 4.1,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR