Ethyl 2-cyclopropyl-6-oxo-1,6-dihydropyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A234161-100MG A234161-250MG
77261-598EA 14.42 USD
77261-598 77261-600
Ethyl 2-cyclopropyl-6-oxo-1,6-dihydropyrimidine-5-carboxylate 97%
Ethyl 2-cyclopropyl-6-oxo-1,6-dihydropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₃
MW: 208.21 g/mol
MDL Number: MFCD16162414
CAS Number: 1044770-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR