6-Bromo-8-methoxyquinoline 97%

Supplier: Ambeed
Warning

A234143-100MG A234143-250MG
77237-950EA 18 USD
77237-950 77746-353
6-Bromo-8-methoxyquinoline 97%
6-Bromo-8-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MDL Number: MFCD11878494
CAS Number: 103028-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR