5,6-Dichloronicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloro-3-pyridinecarboxylic acid, 5,6-Dichloropyridine-3-carboxylic acid, 5,6-Dichloropyridine-3-carboxylicacid

A234088-100G A234088-1G A234088-25G A234088-5G
77288-496EA 162.74 USD
77288-496 77288-498 77288-500 77288-502
5,6-Dichloronicotinic acid 98%
5,6-Dichloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00075181
CAS Number: 41667-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR