6-Fluoro-o-toluic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-methylbenzoic acid

A234011-10G A234011-1G A234011-250MG A234011-25G A234011-5G A234011-100G
77275-812EA 43.26 USD
77275-812 77275-814 77275-816 77275-818 77275-820 77748-402
6-Fluoro-o-toluic acid 97%
6-Fluoro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.1400 g/mol
MDL Number: MFCD06658162
CAS Number: 90259-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR