3,4-Dichloro-2-iodoaniline 95%
Supplier: Ambeed
|
Warning
|
A233249-1G
A233249-250MG
77237-936EA
214.24
USD
77237-936
77237-938
3,4-Dichloro-2-iodoaniline 95%
3,4-Dichloro-2-iodoaniline
Precaución: for Research Use Only
Formula:
C₆H₄Cl₂IN MW: 287.91 g/mol |
MDL Number:
MFCD28128147 CAS Number: 835595-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...