tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 4-((methylamino)methyl)piperidine-1-carboxylate
A233238-10G
A233238-1G
A233238-5G
A233238-25G
77288-468EA
150.38
USD
77288-468
77288-470
77288-472
77748-401
tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate 95%
tert-Butyl-4-((methylamino)methyl)piperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₂₄N₂O₂ MW: 228.3300 g/mol |
MDL Number:
MFCD06656629 CAS Number: 138022-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...