2-Me-D-Trp-OH 95%

Supplier: Ambeed
Warning

A233177-100MG A233177-1G A233177-250MG
77275-782EA 67.98 USD
77275-782 77275-784 77275-786
2-Me-D-Trp-OH 95%
2-Me-D-Trp-OH

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
MW: 218.25 g/mol
MDL Number: MFCD17214441
CAS Number: 140852-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR