(1R,2S,5R)-6-(Benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A233152-100MG
A233152-1G
A233152-250MG
A233152-5G
77261-582EA
21.63
USD
77261-582
77261-584
77261-586
77748-400
(1R,2S,5R)-6-(Benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxylic acid 97%
(1R,2S,5R)-6-(Benzyloxy)-7-oxo-1,6-diazabicyclo[3.2.1]octane-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₆N₂O₄ MW: 276.28 g/mol |
MDL Number:
MFCD28502389 CAS Number: 1174020-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...