1-Boc-4-bromoindole 97%

Supplier: Ambeed
Warning

A232908-1G A232908-250MG A232908-5G
77288-454EA 17 USD
77288-454 77288-456 77288-458
1-Boc-4-bromoindole 97%
1-Boc-4-bromoindole

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrNO₂
MDL Number: MFCD04117874
CAS Number: 676448-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR