1-Cyclopropyl-1H-pyrazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A232846-100MG A232846-1G A232846-250MG A232846-50MG
77261-574EA 42.99 USD
77261-574 77261-576 77261-578 77261-580
1-Cyclopropyl-1H-pyrazole-4-carboxylic acid 97%
1-Cyclopropyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD28405628
CAS Number: 1622883-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR