4-Methoxyphenyl 4-((6-(acryloyloxy)hexyl)oxy)benzoate 97%

Supplier: Ambeed
Warning

A232826-100G A232826-100MG A232826-10G A232826-1G A232826-250MG A232826-25G A232826-5G
77237-922EA 317.24 USD
77237-922 77237-924 77237-926 77237-928 77237-930 77237-932 77237-934
4-Methoxyphenyl 4-((6-(acryloyloxy)hexyl)oxy)benzoate 97%
4-Methoxyphenyl 4-((6-(acryloyloxy)hexyl)oxy)benzoate

Precaución: for Research Use Only

Formula: C₂₃H₂₆O₆
MW: 398.44 g/mol
MDL Number: MFCD27979221
CAS Number: 82200-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR