2-Bromo-1-methyl-1H-imidazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A232802-100MG A232802-1G A232802-250MG
77237-916EA 127.72 USD
77237-916 77237-918 77237-920
2-Bromo-1-methyl-1H-imidazole-4-carboxylic acid 97%
2-Bromo-1-methyl-1H-imidazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
MDL Number: MFCD11100079
CAS Number: 852181-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR