5-Bromo-2,6-di(1H-pyrazol-1-yl)pyrimidin-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: Taminadenant

A232307-1G A232307-250MG
77249-596EA 322 USD
77249-596 77746-350
5-Bromo-2,6-di(1H-pyrazol-1-yl)pyrimidin-4-amine 97%
5-Bromo-2,6-di(1H-pyrazol-1-yl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₀H₈BrN₇
MDL Number: MFCD30489281
CAS Number: 1337962-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR