RQ-002303078 98+%

Supplier: Ambeed
Warning

Synonyms: RQ-00203078, 4-(N-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)-N-(4-(trifluoromethoxy)benzyl)sulphamoyl)benzoic acid

A231850-10MG A231850-1MG A231850-50MG A231850-5MG
77237-890EA 47.38 USD
77237-890 77237-892 77237-894 77237-896
RQ-002303078 98+%
RQ-002303078

Precaución: for Research Use Only

Formula: C₂₁H₁₃ClF₆N₂O₅S
MW: 554.85 g/mol
MDL Number: MFCD29049510
CAS Number: 1254205-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR