(4-Amino-2-methyl-5-pyrimidinyl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: Toxopyrimidine, 4-Amino-2-methyl-5-pyrimidinemethanol, 4-Amino-5-hydroxymethyl-2-methylpyrimidine

A231683-1G A231683-5G
77288-410EA 100.94 USD
77288-410 77288-412
(4-Amino-2-methyl-5-pyrimidinyl)methanol 95%
(4-Amino-2-methyl-5-pyrimidinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MW: 139.1552 g/mol
MDL Number: MFCD00849239
CAS Number: 73-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR