2-Methoxy-5-methylbenzaldehyde 95%

Supplier: Ambeed
Warning

A231525-100MG A231525-1G A231525-250MG A231525-25G A231525-5G
77261-552EA 12 USD
77261-552 77261-554 77261-556 77261-558 77261-560
2-Methoxy-5-methylbenzaldehyde 95%
2-Methoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD02261770
CAS Number: 7083-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR