[2,2'-Bipyridine]-5,5'-diyldimethanol 96%

Supplier: Ambeed
Warning

A231384-1G A231384-250MG
77237-886EA 76 USD
77237-886 77237-888
[2,2'-Bipyridine]-5,5'-diyldimethanol 96%
[2,2'-Bipyridine]-5,5'-diyldimethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD28405910
CAS Number: 63361-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR