4-Chloro-2-hydrazinylpyridine 95%

Supplier: Ambeed
Warning

A230942-1G A230942-250MG
77237-850EA 94.76 USD
77237-850 77237-852
4-Chloro-2-hydrazinylpyridine 95%
4-Chloro-2-hydrazinylpyridine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.57 g/mol
MDL Number: MFCD10000027
CAS Number: 364757-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR