4,6-Difluoroindole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A230931-1G A230931-250MG A230931-5G
77275-736EA 23 USD
77275-736 77275-738 77275-740
4,6-Difluoroindole-2-carboxylic acid 97%
4,6-Difluoroindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅F₂NO₂
MDL Number: MFCD01863158
CAS Number: 247564-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR