(R)-4-Propyldihydrofuran-2(3H)-one 97%

Supplier: Ambeed
Warning

A230886-1G A230886-250MG A230886-5G
77237-844EA 85 USD
77237-844 77237-846 77237-848
(R)-4-Propyldihydrofuran-2(3H)-one 97%
(R)-4-Propyldihydrofuran-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MDL Number: MFCD29905061
CAS Number: 63095-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR