4-Chlorophenyl cyclopropyl ketone 98%

Supplier: Ambeed
Warning

A230740-100G A230740-25G A230740-5G
77288-364EA 255 USD
77288-364 77288-366 77288-368
4-Chlorophenyl cyclopropyl ketone 98%
4-Chlorophenyl cyclopropyl ketone

Precaución: for Research Use Only

Formula: C₁₀H₉ClO
MDL Number: MFCD00001295
CAS Number: 6640-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR