(R,E)-4-((tert-Butyldimethylsilyl)oxy)pent-2-enoic acid 97%

Supplier: Ambeed
Warning

A230730-1G A230730-250MG
77261-540EA 55.62 USD
77261-540 77261-542
(R,E)-4-((tert-Butyldimethylsilyl)oxy)pent-2-enoic acid 97%
(R,E)-4-((tert-Butyldimethylsilyl)oxy)pent-2-enoic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₂O₃Si
MW: 230.38 g/mol
MDL Number: MFCD31381098
CAS Number: 2126144-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR