4-Fluoro-L-2-phenylglycine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-L-phenylglycine, (S)-2-Amino-2-(4-fluorophenyl)acetic Acid, H-Phg(4-F)-OH, (S)-4-Fluorophenylglycine

A230536-100G A230536-10G A230536-1G A230536-25G A230536-5G
77249-558EA 131 USD
77249-558 77249-560 77249-562 77249-564 77249-566
4-Fluoro-L-2-phenylglycine 97%
4-Fluoro-L-2-phenylglycine

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MDL Number: MFCD00213799
CAS Number: 19883-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR