Ethyl 4-(aminomethyl)benzoate hydrochloride 97%

Supplier: Ambeed
Warning

A230197-1G A230197-250MG A230197-25G A230197-5G
77237-836EA 6 USD
77237-836 77237-838 77237-840 77237-842
Ethyl 4-(aminomethyl)benzoate hydrochloride 97%
Ethyl 4-(aminomethyl)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD00859552
CAS Number: 6232-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR