N-(p-Toluenesulfonyl)glycine 97%

Supplier: Ambeed
Warning

Synonyms: N-Tosylglycine, N-p-Tosylglycine, Tos-Gly-OH

A230047-10G A230047-25G A230047-5G A230047-100G
77249-542EA 21 USD
77249-542 77249-544 77249-546 77746-343
N-(p-Toluenesulfonyl)glycine 97%
N-(p-Toluenesulfonyl)glycine

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄S
MW: 229.25 g/mol
MDL Number: MFCD00045898
CAS Number: 1080-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR