2,5-Dimethoxythiophenol 98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Dimethoxybenzenethiol, 2,5-Dimethoxyphenyl mercaptan, 1,4-Dimethoxy-2-mercaptobenzene, 2,5-Dimethoxybenzene-1-thiol

A230011-1G A230011-25G A230011-5G
77249-536EA 30.9 USD
77249-536 77249-538 77249-540
2,5-Dimethoxythiophenol 98%
2,5-Dimethoxythiophenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
MDL Number: MFCD00052738
CAS Number: 1483-27-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR