4-Acetoxy-3-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-Formyl-2-methoxyphenyl acetate, Vanillin acetate

A229975-100G A229975-25G A229975-500G
77288-354EA 55 USD
77288-354 77288-356 77288-358 77748-389
4-Acetoxy-3-methoxybenzaldehyde 98%
4-Acetoxy-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00003362
CAS Number: 881-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR