Bis(4-nitrophenyl)amine 96%

Supplier: Ambeed
Danger

Synonyms: 4,4'-Dinitrodiphenylamine

A229887-100MG A229887-10G A229887-1G A229887-250MG A229887-5G
77288-344EA 10 USD
77288-344 77288-346 77288-348 77288-350 77288-352
Bis(4-nitrophenyl)amine 96%
Bis(4-nitrophenyl)amine

Precaución: for Research Use Only

Formula: C₁₂H₉N₃O₄
MDL Number: MFCD00437044
CAS Number: 1821-27-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR