4'-Bromopropiophenone 98%

Supplier: Ambeed
Warning

A229708-100G A229708-25G
77288-336EA 10 USD
77288-336 77288-338 77288-340 77288-342
4'-Bromopropiophenone 98%
4'-Bromopropiophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD00000106
CAS Number: 10342-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR