3-Amino-6-thiophen-2-yl-4-trifluoromethyl-thieno[2,3-b]pyridine-2-carboxylic acid (4-fluoro-phenyl)-amide 97%

Supplier: Ambeed
Warning

Synonyms: FDI-6

A229685-10MG A229685-1MG A229685-5MG
77237-826EA 129.78 USD
77237-826 77237-828 77237-830
3-Amino-6-thiophen-2-yl-4-trifluoromethyl-thieno[2,3-b]pyridine-2-carboxylic acid (4-fluoro-phenyl)-amide 97%
3-Amino-6-thiophen-2-yl-4-trifluoromethyl-thieno[2,3-b]pyridine-2-carboxylic acid (4-fluoro-phenyl)-amide

Precaución: for Research Use Only

Formula: C₁₉H₁₁F₄N₃OS₂
MW: 437.43 g/mol
MDL Number: MFCD01416826
CAS Number: 313380-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR