2-Aminothiazole-5-carbonitrile 97%

Supplier: Ambeed
Danger

A229383-1G A229383-25G A229383-5G
77261-532EA 43.26 USD
77261-532 77261-534 77261-536
2-Aminothiazole-5-carbonitrile 97%
2-Aminothiazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₄H₃N₃S
MW: 125.20 g/mol
MDL Number: MFCD08275724
CAS Number: 51640-52-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR