4-(Trifluoromethyl)benzophenone 98%

Supplier: Ambeed
Warning

Synonyms: Phenyl(4-(trifluoromethyl)phenyl)methanone

A228892-1G A228892-25G A228892-5G
77288-322EA 14 USD
77288-322 77288-324 77288-326
4-(Trifluoromethyl)benzophenone 98%
4-(Trifluoromethyl)benzophenone

Precaución: for Research Use Only

Formula: C₁₄H₉F₃O
MDL Number: MFCD00000400
CAS Number: 728-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR