3-Cyanobenzoic acid 98%

Supplier: Ambeed
Warning

A228622-100G A228622-10G A228622-25G A228622-5G
77237-812EA 42.23 USD
77237-812 77237-814 77237-816 77237-818
3-Cyanobenzoic acid 98%
3-Cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₂
MW: 147.13 g/mol
MDL Number: MFCD00002486
CAS Number: 1877-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR