3-Phenyl-1H-pyrazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3-Phenyl-1H-pyrazole-4-carboxaldehyde

A228546-10G A228546-1G A228546-250MG A228546-25G A228546-5G
77288-312EA 120.51 USD
77288-312 77288-314 77288-316 77288-318 77288-320
3-Phenyl-1H-pyrazole-4-carbaldehyde 97%
3-Phenyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MW: 172.187 g/mol
MDL Number: MFCD00216971
CAS Number: 26033-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR