5-Fluorothiazolo[4,5-b]pyridin-2-amine 97%

Supplier: Ambeed
Warning

A228450-250MG A228450-100MG
77237-798EA 154.5 USD
77237-798 77746-339
5-Fluorothiazolo[4,5-b]pyridin-2-amine 97%
5-Fluorothiazolo[4,5-b]pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₄FN₃S
MW: 169.18 g/mol
MDL Number: MFCD14581837
CAS Number: 1206248-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR