(Tetrahydrofuran-2-yl)acetonitrile 98%

Supplier: Ambeed
Warning

A228410-100MG A228410-250MG
77249-518EA 29 USD
77249-518 77249-520
(Tetrahydrofuran-2-yl)acetonitrile 98%
(Tetrahydrofuran-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₆H₉NO
MDL Number: MFCD03206782
CAS Number: 33414-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR