2-Fluoro-DL-phenylalanine 97%

Supplier: Ambeed
Danger

A228325-1G A228325-25G A228325-5G
77249-512EA 19 USD
77249-512 77249-514 77249-516
2-Fluoro-DL-phenylalanine 97%
2-Fluoro-DL-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MDL Number: MFCD00004271
CAS Number: 2629-55-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR