3-(3-Methoxyphenoxy)propionic acid 95%

Supplier: Ambeed
Warning

A228289-1G A228289-250MG A228289-5G
77275-696EA 38 USD
77275-696 77275-698 77275-700
3-(3-Methoxyphenoxy)propionic acid 95%
3-(3-Methoxyphenoxy)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00045943
CAS Number: 49855-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR