5-Bromo-2-methylindole 98%
Supplier: Ambeed
|
Warning
|
A228276-100MG
A228276-1G
A228276-250MG
A228276-25G
A228276-5G
A228276-10G
77261-504EA
5
USD
77261-504
77261-506
77261-508
77261-510
77261-512
77748-384
5-Bromo-2-methylindole 98%
5-Bromo-2-methylindole
Precaución: for Research Use Only
Formula:
C₉H₈BrN |
MDL Number:
MFCD01863677 CAS Number: 1075-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...